Assay Details
| Gene Symbol: SLC39A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098132.1 | NP_001091601.1 | 7 - 8 | 842 | 61 | |
| GenBank mRNA | BC140552.1 | - | 7 - 8 | 842 | 61 | |
More Information
Set Membership: |
|

SLC39A13
Genomic Map