Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098143.2 | NP_001091612.1 | 1 - 2 | 61 | 111 | |
| GenBank mRNA | BC140643.1 | - | 1 - 2 | 62 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC140643.1, NM_001098143.2 transcripts.
Set Membership: |
|

SEPTIN12
Genomic Map