Assay Details
| Gene Symbol: SEPTIN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098143.2 | NP_001091612.1 | 8 - 9 | 911 | 86 | |
| GenBank mRNA | BC140643.1 | - | 8 - 9 | 912 | 86 | |
More Information
Set Membership: |
|

SEPTIN12
Genomic Map