Assay Details
| Gene Symbol: ZFP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098146.1 | NP_001091615.1 | 1 - 2 | 28 | 86 | |
| GenBank mRNA | BC140589.1 | - | 1 - 2 | 28 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC140589.1, NM_001098146.1 transcripts.
Set Membership: |
|

ZFP2
Genomic Map