Assay Details
| Gene Symbol: GNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099033.1 | NP_001092503.1 | 8 - 9 | 827 | 65 | |
| GenBank mRNA | BC142449.1 | - | 8 - 9 | 827 | 65 | |
More Information
Set Membership: |
|

GNB4
Genomic Map