Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099038.1 | NP_001092508.1 | 5 - 6 | 616 | 69 | |
| GenBank mRNA | BC142195.1 | - | 5 - 6 | 616 | 69 | |
More Information
Set Membership: |
|

MAP2K4
Genomic Map