Assay Details
| Gene Symbol: PRSS54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099044.2 | NP_001092514.1 | 2 - 3 | 400 | 59 | |
| GenBank mRNA | BC142059.1 | - | 2 - 3 | 401 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC142059.1, NM_001099044.2 transcripts.
Set Membership: |
|

PRSS54
Genomic Map