Assay Details
Gene Symbol: ANKRD55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099057.2 | NP_001092527.1 | 9 - 10 | 1608 | 79 | |
GenBank mRNA | BC142446.1 | - | 9 - 10 | 1609 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
