Assay Details
| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099085.2 | NP_001092555.1 | 4 - 5 | 1594 | 64 | |
| GenBank mRNA | BC142321.1 | - | 4 - 5 | 1595 | 64 | |
More Information
Set Membership: |
|

EEF1AKNMT
Genomic Map