Assay Details
| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099085.2 | NP_001092555.1 | 7 - 8 | 2115 | 68 | |
| GenBank mRNA | BC142321.1 | - | 7 - 8 | 2116 | 68 | |
More Information
Set Membership: |
|

EEF1AKNMT
Genomic Map