Assay Details
| Gene Symbol: SLC1A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099090.1 | NP_001092560.1 | 7 - 8 | 1330 | 71 | |
| GenBank mRNA | BC146139.1 | - | 7 - 8 | 1330 | 71 | |
More Information
Set Membership: |
|

SLC1A6
Genomic Map