Assay Details
| Gene Symbol: AREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099092.1 | NP_001092562.1 | 2 - 3 | 522 | 76 | |
| GenBank mRNA | BC142081.1 | - | 2 - 3 | 522 | 76 | |
More Information
Set Membership: |
|

AREG
Genomic Map