Assay Details
| Gene Symbol: RCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099097.1 | NP_001092567.1 | 6 - 7 | 711 | 67 | |
| GenBank mRNA | BC146167.1 | - | 6 - 7 | 711 | 67 | |
More Information
Set Membership: |
|

RCE1
Genomic Map