Assay Details
| Gene Symbol: POGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099100.1 | NP_001092570.1 | 4 - 5 | 470 | 71 | |
| GenBank mRNA | BC142233.1 | - | 4 - 5 | 470 | 71 | |
More Information
Set Membership: |
|

POGK
Genomic Map