Assay Details
| Gene Symbol: KCNH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099101.1 | NP_001092571.1 | 2 - 3 | 330 | 60 | |
| GenBank mRNA | BC142496.1 | - | 2 - 3 | 330 | 60 | |
More Information
Set Membership: |
|

KCNH2
Genomic Map