Assay Details
| Gene Symbol: RND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191158.3 | NP_001178087.1 | 3 - 4 | 591 | 72 | |
| GenBank mRNA | BC142513.1 | - | 3 - 4 | 592 | 72 | |
More Information
Set Membership: |
|

RND3
Genomic Map