Assay Details
| Gene Symbol: PIMREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099109.1 | NP_001092579.1 | 1 - 2 | 337 | 128 | |
| GenBank mRNA | BC142479.1 | - | 1 - 2 | 337 | 128 | |
More Information
Set Membership: |
|

PIMREG
Genomic Map