Assay Details
| Gene Symbol: PPM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099118.2 | NP_001092588.1 | 3 - 4 | 818 | 65 | |
| GenBank mRNA | BC142293.1 | - | 3 - 4 | 819 | 65 | |
More Information
Set Membership: |
|

PPM1L
Genomic Map