Assay Details
| Gene Symbol: PLLP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099123.1 | NP_001092593.1 | 2 - 3 | 440 | 95 | |
| GenBank mRNA | BC146177.1 | - | 2 - 3 | 440 | 95 | |
More Information
Set Membership: |
|

PLLP
Genomic Map