Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099129.1 | NP_001092599.1 | 1 - 2 | 176 | 110 | |
| GenBank mRNA | BC146199.1 | - | 1 - 2 | 176 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146199.1, NM_001099129.1 transcripts.
Set Membership: |
|

CEP19
Genomic Map