Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099129.1 | NP_001092599.1 | 2 - 3 | 373 | 85 | |
| GenBank mRNA | BC146199.1 | - | 2 - 3 | 373 | 85 | |
More Information
Set Membership: |
|

CEP19
Genomic Map