Assay Details
| Gene Symbol: EMC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099143.2 | NP_001092613.1 | 4 - 5 | 590 | 80 | |
| GenBank mRNA | BC134707.1 | - | 4 - 5 | 599 | 80 | |
| BC142029.1 | - | 4 - 5 | 591 | 80 | ||
More Information
Set Membership: |
|

EMC7
Genomic Map