Assay Details
| Gene Symbol: CCL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099162.2 | NP_001092632.1 | 2 - 3 | 234 | 88 | |
| GenBank mRNA | BC142465.1 | - | 2 - 3 | 235 | 88 | |
More Information
Set Membership: |
|

CCL22
Genomic Map