Assay Details
| Gene Symbol: IGFLR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099167.2 | NP_001092637.1 | 3 - 4 | 596 | 83 | |
| GenBank mRNA | BC142051.1 | - | 3 - 4 | 597 | 83 | |
More Information
Set Membership: |
|

IGFLR1
Genomic Map