Assay Details
| Gene Symbol: SYT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099171.2 | NP_001092641.1 | 1 - 2 | 231 | 70 | |
| GenBank mRNA | BC146251.1 | - | 1 - 2 | 232 | 70 | |
More Information
Set Membership: |
|

SYT11
Genomic Map