Assay Details
| Gene Symbol: SDR16C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099707.1 | NP_001093177.1 | 3 - 4 | 506 | 96 | |
| GenBank mRNA | BC142138.1 | - | 3 - 4 | 506 | 96 | |
More Information
Set Membership: |
|

SDR16C6
Genomic Map