Assay Details
| Gene Symbol: STX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099719.1 | NP_001093189.1 | 5 - 6 | 506 | 62 | |
| GenBank mRNA | BC142214.1 | - | 5 - 6 | 506 | 62 | |
More Information
Set Membership: |
|

STX18
Genomic Map