Assay Details
| Gene Symbol: ENO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101125.1 | NP_001094595.1 | 5 - 6 | 471 | 64 | |
| GenBank mRNA | BC150078.1 | - | 5 - 6 | 471 | 64 | |
More Information
Set Membership: |
|

ENO2
Genomic Map