Assay Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101129.1 | NP_001094599.1 | 1 - 2 | 196 | 77 | |
| GenBank mRNA | BC150111.1 | - | 1 - 2 | 196 | 77 | |
More Information
Set Membership: |
|

RAB6B
Genomic Map