Assay Details
| Gene Symbol: SLC9A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101146.1 | NP_001094616.1 | 14 - 15 | 1743 | 65 | |
| GenBank mRNA | BC151331.1 | - | 14 - 15 | 1743 | 65 | |
More Information
Set Membership: |
|

SLC9A6
Genomic Map