Assay Details
| Gene Symbol: NABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101151.1 | NP_001094621.1 | 3 - 4 | 404 | 69 | |
| GenBank mRNA | BC147994.1 | - | 3 - 4 | 404 | 69 | |
More Information
Set Membership: |
|

NABP2
Genomic Map