Assay Details
| Gene Symbol: ALAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101154.1 | NP_001094624.1 | 10 - 11 | 1955 | 66 | |
| GenBank mRNA | BC147978.1 | - | 10 - 11 | 1955 | 66 | |
More Information
Set Membership: |
|

ALAS1
Genomic Map