Assay Details
| Gene Symbol: CNOT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101161.1 | NP_001094631.1 | 3 - 4 | 357 | 85 | |
| GenBank mRNA | BC150116.1 | - | 3 - 4 | 357 | 85 | |
More Information
Set Membership: |
|

CNOT6
Genomic Map