Assay Details
| Gene Symbol: PEX26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101172.1 | NP_001094642.1 | 2 - 3 | 354 | 77 | |
| GenBank mRNA | BC149365.1 | - | 2 - 3 | 354 | 77 | |
More Information
Set Membership: |
|

PEX26
Genomic Map