Assay Details
| Gene Symbol: RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101192.2 | NP_001094662.1 | 2 - 3 | 244 | 74 | |
| GenBank mRNA | BC151528.1 | - | 2 - 3 | 245 | 74 | |
More Information
Set Membership: |
|

RBP5
Genomic Map