Assay Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101200.2 | NP_001094670.1 | 3 - 4 | 320 | 63 | |
| GenBank mRNA | BC151255.1 | - | 3 - 4 | 321 | 63 | |
More Information
Set Membership: |
|

TAF8
Genomic Map