Assay Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101204.1 | NP_001094674.1 | 6 - 7 | 1577 | 55 | |
| GenBank mRNA | BC150109.1 | - | 6 - 7 | 1577 | 55 | |
More Information
Set Membership: |
|

DGCR8
Genomic Map