Assay Details
| Gene Symbol: WEE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101205.1 | NP_001094675.1 | 2 - 3 | 336 | 70 | |
| GenBank mRNA | BC151481.1 | - | 2 - 3 | 795 | 70 | |
| BC151685.1 | - | 2 - 3 | 336 | 70 | ||
More Information
Set Membership: |
|

WEE1
Genomic Map