Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101218.1 | NP_001094688.1 | 5 - 6 | 941 | 97 | |
| GenBank mRNA | BC150130.1 | - | 5 - 6 | 941 | 97 | |
More Information
Set Membership: |
|

ZNF131
Genomic Map