Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101225.2 | NP_001094695.1 | 1 - 2 | 246 | 71 | |
| GenBank mRNA | BC149592.1 | - | 1 - 2 | 247 | 71 | |
More Information
Set Membership: |
|

POLE4
Genomic Map