Assay Details
| Gene Symbol: ZFP36L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101234.1 | NP_001094704.1 | 1 - 2 | 193 | 67 | |
| GenBank mRNA | BC151440.1 | - | 1 - 2 | 193 | 67 | |
More Information
Set Membership: |
|

ZFP36L1
Genomic Map