Assay Details
| Gene Symbol: AKIRIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101236.1 | NP_001094706.1 | 3 - 4 | 609 | 100 | |
| GenBank mRNA | BC151342.1 | - | 3 - 4 | 609 | 100 | |
More Information
Set Membership: |
|

AKIRIN1
Genomic Map