Assay Details
| Gene Symbol: BTBD19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101241.1 | NP_001094711.1 | 7 - 8 | 1019 | 68 | |
| GenBank mRNA | BC149224.1 | - | 7 - 8 | 1019 | 68 | |
More Information
Set Membership: |
|

BTBD19
Genomic Map