Assay Details
| Gene Symbol: EFNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101249.2 | NP_001094719.1 | 2 - 3 | 491 | 73 | |
| GenBank mRNA | BC148126.1 | - | 2 - 3 | 492 | 73 | |
More Information
Set Membership: |
|

EFNA4
Genomic Map