Assay Details
| Gene Symbol: RBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101259.2 | NP_001094729.1 | 2 - 3 | 327 | 87 | |
| GenBank mRNA | BC151542.1 | - | 2 - 3 | 329 | 87 | |
More Information
Set Membership: |
|

RBP2
Genomic Map