Assay Details
| Gene Symbol: MARVELD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101262.1 | NP_001094732.1 | 1 - 2 | 2277 | 76 | |
| GenBank mRNA | BC151267.1 | - | 1 - 2 | 2277 | 76 | |
More Information
Set Membership: |
|

MARVELD1
Genomic Map