Assay Details
| Gene Symbol: SHISA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101265.1 | NP_001094735.1 | 1 - 2 | 477 | 72 | |
| GenBank mRNA | BC149221.1 | - | 1 - 2 | 477 | 72 | |
More Information
Set Membership: |
|

SHISA2
Genomic Map