Assay Details
| Gene Symbol: BARX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101266.1 | NP_001094736.1 | 3 - 4 | 849 | 65 | |
| GenBank mRNA | BC148101.1 | - | 3 - 4 | 849 | 65 | |
More Information
Set Membership: |
|

BARX2
Genomic Map