Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101269.2 | NP_001094739.1 | 2 - 3 | 233 | 63 | |
| GenBank mRNA | BC151623.1 | - | 2 - 3 | 234 | 63 | |
More Information
Set Membership: |
|

NRSN2
Genomic Map