Assay Details
| Gene Symbol: ITLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101275.1 | NP_001094745.1 | 1 - 2 | 67 | 93 | |
| GenBank mRNA | AM749043.1 | - | 1 - 2 | 67 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AM749043.1, NM_001101275.1 transcripts.
Set Membership: |
|

ITLN2
Genomic Map